Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 1110059E24Rik Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569956 Supplier Invitrogen™ Supplier No. PA569956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 85%; Rat: 100%.

1110059E24Rik is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 1110059E24Rik
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene 1110059E24Rik
Gene Accession No. Q9CQ90
Gene Alias 1110059E24Rik; RIKEN cDNA 1110059E24 gene; uncharacterized protein C9orf85 homolog
Gene Symbols 1110059E24Rik
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human 1110059E24Rik (aa 21-70).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66206
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.