Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIPK2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570131 Supplier Thermo Scientific Supplier No. PA570131
Add to Cart
Add to Cart

DIPK2A Polyclonal Antibody

DIPK2A Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen DIPK2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DIPK2A
Gene Accession No. Q3USZ8
Gene Alias 1190002N15Rik; Ab2-095; C3orf58; cc058; chromosome 3 open reading frame 58; deleted in autism 1; deleted in autism protein 1; Deleted in autism protein 1 homolog; DIA1; Dipk2a; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; hypoxia and Akt induced stem cell factor; hypoxia and AKT-induced stem cell factor; RGD1309079; RIKEN cDNA 1190002N15 gene; similar to Ab2-095; UPF0672 protein C3orf58; UPF0672 protein C3orf58 homolog
Gene Symbols DIPK2A
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of mouse 1190002N15Rik.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68861
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.