Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
12-Lipoxygenase Antibody (SR2151), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | 12-Lipoxygenase |
---|---|
Clone | SR2151 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
12-Lipoxygenase Monoclonal antibody specifically detects 12-Lipoxygenase in Human, Rat samples. It is validated for Western BlotSpecifications
12-Lipoxygenase | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
12(S)-lipoxygenase, 12-LOX, 12S-lipoxygenase, 12S-LOXarachidonate 12-lipoxygenase, 12S-type, arachidonate 12-lipoxygenase, EC 1.13.11, EC 1.13.11.31, LOG12, platelet 12-LOX, platelet-type 12-lipoxygenase, Platelet-type lipoxygenase 12 | |
A synthesized peptide derived from human 12-Lipoxygenase (Uniprot #: P18054) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2151 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
239 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title