Learn More
12-Mercaptododecylphosphonic acid, 95%
Description
Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Application to the Extraction of Heavy Metals. Recent works using phosphonic acid PA) molecules have demonstrated the ability of 12-mercaptododecylphosphonic acid (MDPA)to strengthen the bond between Au and TiO 2 , leading to an increase in h K.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 94°C to 96°C |
| Linear Formula | HS(CH2)12P(O)(OH)2 |
| Quantity | 250 mg |
| Sensitivity | Air Sensitive |
| Formula Weight | 282.38 |
| Percent Purity | 95% |
| Chemical Name or Material | 12-Mercaptododecylphosphonic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.