Learn More
12-Molybdophosphoric acid hydrate, ACS
Description
12-Molybdophosphoric acid hydrate is used as a precursor to hybrid organic-inorganic based molecular batteries with application as cation-insertion electrode. It finds application as an analytical reagent in thin layer chromatography for staining phenolics, hydrocarbon waxes, alkaloids and steroids. It acts as a reactant for the preparation of catalysts for the nitration of aromatic compounds, highly dispersed MoOx/CNTs (carbon nanotubes) as a support for Pt catalysts and anchored Wilkinson catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 78°C to 90°C |
| Color | Yellow |
| Density | 3.15 |
| Odor | Odorless |
| Quantity | 100 g |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in water,ethanol and ether. |
| Formula Weight | 1825.25 (Anhydrous) |
| Grade | ACS |
| Physical Form | Granules |
| Show More |
Safety and Handling
| Hazard Category | H272-H290-H314-H335-H373 |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c |
| DOTInformation | DOT Class: 8; Packaging Group: II |
| EINECSNumber | 234-713-5 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.