Learn More
12-Tungstophosphate hydrate, Reagent Grade
Description
12-Tungstophosphate hydrate is used in histology as a component for staining of cell specimens. It binds to fibrin, collagen and fibers of connective tissues. It is also used to replace the anions of dyes from these materials. In solution, it acts as a homogeneous catalyst and involved in the hydrolysis of propene to give 2-propanol. It is supported with alumina and silica and act as a heterogeneous catalyst involved in the dehydration of 2-propanol to propene and methanol to hydrocarbons. In analytical chemistry, it is used to precipitate proteins and termed as universal precipitant for polar proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 95°C |
| Color | White through Yellow |
| Physical Form | Powder |
| Quantity | 50 g |
| UN Number | UN3260 |
| Solubility Information | Soluble in water,dichloromethane,acetonitrile,acetic acid and ionic liquids. |
| IUPAC Name | phosphoric acid dodecakis(trioxotungsten) hydrate |
| Formula Weight | 2880.20 (Anhydrous) |
| Grade | Reagent |
| Sensitivity | Moisture & Light Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H314-H335 |
| Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.