Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiTrap™ Capto™ Octyl HIC Column

Catalog No. 12101329 Shop All Cytiva Products
Change view
Click to view available options
Capacity (Metric):
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
12-101-329 5 mL
12-101-330 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-101-329 Supplier Cytiva Supplier No. 17546512
Add to Cart
Add to Cart

Prepacked with Capto™ Octyl hydrophobic interaction chromatography (HIC) resin for small scale purification.

HiTrap™ Capto™ Octyl column is prepacked with a HIC resin for small scale purification

  • Prepacked with Capto™ Octyl high-flow and high-capacity HIC resin for capture and intermediate purification processes
  • Lab-scale column format, suitable for screening of selectivity, binding and elution conditions, and for small-scale purifications in research and process development
  • Convenient HiTrap™ format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA™ system
  • Manufactured from biocompatible polypropylene that does not interact with biomolecules
  • Has an octyl ligand for the capture and intermediate purification of monoclonal antibodies (mAbs), mAb variants, and other biomolecules
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Product Type HiTrap Octyl FF
Flow Rate 5 mL/min.
Quantity 5/Pk.
Ligand Type Octyl
Max. Pressure 3 bar
Particle Size 75 μm
pH Range 3 to 13
Content And Storage 4°C to 30°C
Storage Buffer 20% Ethanol
Binding Capacity 25 mg
Height (Metric) Bed 25 mm
Capacity (Metric) 5 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.