Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bromoacetaldehyde diethyl acetal

Catalog No. 123986100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-398-6100G 100 g
12-398-61KG 1 kg
12-398-6500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 12-398-6100G Supplier Sigma Aldrich Supplier No. 123986100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • May darken in storagebromoacetaldehyde diethyl acetal has been used in the synthesis of monomer 2-(2,2-dimethoxy)ethoxyethyl vinyl ether
  • It has been used as synthetic building block

Specifications

CAS 2032-35-1
Density 1.31 g/mL (at 25°C (literature))
Boiling Point 66°C to 67°C (18 mmHg)
Molecular Formula C6H13BrO2
Refractive Index n20/D 1.439 (literature)
Linear Formula BrCH2CH(OC2H5)2
MDL Number MFCD00000214
Quantity 100 g
Synonym 2-Bromo-1,1-diethoxyethane; Bromoacetal
Molecular Weight (g/mol) 197.07
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.