Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Superslip™ Cover Slips
Shop All Fisherbrand Products
Click to view available options
Thickness (Metric):
0.13 to 0.17 mm (#1)
0.16 to 0.19 mm (#1.5)
Dimensions (L x W):
40 x 24 mm
50 x 24 mm
60 x 24 mm
Description
All Superslip™ Cover Slips feature a high-spectral transmission quality and finely adaptive refractive index suited for microscopy.
- Uniquely developed to reduce sticking
- Can be used with automated coverslippers or for manual use in humid environments
- Built-in moisture resistance
- Manufactured with colorless borosilicate glass
- Available in a variety of sizes and thicknesses
Specifications
Specifications
| Thickness (Metric) | 0.13 to 0.17 mm (#1) |
| Dimensions (L x W) | 40 x 24 mm |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction