Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ 96-Well Polypropylene DeepWell™ Sample Processing & Storage Plates with Shared-Wall Technology
GSA_VAGREENER_CHOICE

Catalog No. 12565606
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Well Shape:
Round
Square, Conical Bottom (V-well)
Volume (Metric) Well:
1300 μL
2000 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility Well Shape Volume (Metric) Well
12-565-606 Non-sterile Round 2000 μL
12-565-395 Sterile Round 1300 μL
12-565-394 Non-sterile Round 1300 μL
12-565-605 Sterile Round 2000 μL
12565650 Non-sterile Square, Conical Bottom (V-well) 2000 μL
12565651 Sterile Square, Conical Bottom (V-well) 2000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 12-565-606 Supplier Thermo Scientific™ Supplier No. 278752
Add to Cart
Add to Cart

Optimize storage capacity and improve mixing using these DeepWell™ storage plates featuring shared-wall technology. Shared-Wall reduces the height of the plate, increases the diameter of the well for sample extraction.

Optimize storage capacity and improve mixing using Thermo Scientific™ Nunc™ 1.3 and 2.0mL DeepWell™ Plates with Shared-Wall Technology. Ideal for sample collection, storage, combinatorial chemistry and library applications, and resistant to most chemicals, solvents and alcohols used in combinatorial chemistry.

The large well diameters offer ideal access and excellent mixing characteristics, and the U shaped well enhances compatibility with the magnetic bead extraction. The V shaped well bottom ensures low dead-volume and high sample recovery during extraction

All Thermo Scientific plates are manufactured in our cleanroom facility or clean environment to ensure molecular grade quality. Our wide range of polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

Features:

  • Ideal as a collection plate for Nunc Filter Plates
  • Storage plate for compounds, samples or biomolecules
  • Round-bottom wells reduce liquid retention
  • Round well enhances mixing
  • Automated molecular diagnostic workflows
  • Magnetic Bead Extraction compatible
  • Ideal for Kitting

Working volumes:

  • 260251 and 260252 50-1000 μL/well
  • 278743 and 278752 50-1900 μL/well

Recommended for:

Sample collection; storage; combinatorial chemistry; drug library applications; bacterial and yeast growth; collection plates for Nunc filter plates; NGS and genomic library prep

Compatible Sealing: Adhesive and Heat seals
Hazardous: No
Max Centrifugal Force: 2000 g

All plates are ANSI format for compatibility with automated systems.

To further assist in sample storage and tracking, all plates can be customized with non-repeating barcoding and labelling. OEM options for custom logos, colors and full packaging support are available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Custom Barcoding Available
Certifications/Compliance Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Format Standard
Lid Without Lid
Description 96-well Polypropylene Sample Processing & Storage Plates
Array 8 x 12
Autoclavable Autoclavable
Packaging 60/Case
Binding Type Low binding PP
Product Line Nunc
Color Natural
Product Type 96-well plate
For Use With (Equipment) ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
Material Polypropylene
Volume (Metric) Well 2000 μL
No. of Wells 96
Volume (Metric) Working 1700 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
Dnase Free Yes
For Use With (Application) Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case 60
No. per Pack 5
Quantity 60/Cs.
Regulatory Status For Research Use Only. Not for use in diagnostic procedures.
RNase Free Yes
Fit for Purpose PCR Ready
Plate Size ANSI/ SLAS Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.