Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl Methanesulfonate

Catalog No. 12992525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-992-525G 25 g
12-992-5100G 100 g
12-992-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 12-992-525G Supplier Sigma Aldrich Supplier No. 12992525G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl Methanesulfonate

Specifications

CAS 66-27-3
Density 1.3 g/mL at 25°C
Boiling Point 202°C to 203°C
Molecular Formula C2H6O3S
Refractive Index n20/D 1.414
Linear Formula CH3SO3CH3
MDL Number MFCD00007557
Quantity 25 g
Synonym Methanesulfonic acid methyl ester
Molecular Weight (g/mol) 110.13 g/mol
Biological Activity Lungs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.