Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
12D10-Fab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA541773

This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
12D10-Fab Recombinant Monoclonal antibody specifically detects 12D10-Fab in Chemical samples. It is validated for ELISA
Specifications
12D10-Fab | |
Recombinant Monoclonal | |
Unconjugated | |
12D10 Fab; 12D10-Fab; F7 | |
Human F7 | |
100 μg | |
Primary | |
Antibody | |
Chemical |
ELISA | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein L | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction