Learn More
Description
- The Non-species specific 12S-HETE ELISA Kit (Colorimetric) measures non-species specific 12S-HETE in culture supernatants and plasma from any species. cited sample types include serum, renal interstitial fluid and tissue.
- Standard Curve Range: 195 - 50,000 pg/mL
- Sensitivity: 146 pg/mL
- Assay Type: Competitive ELISA
- Inter-Assay: Mean 11.3%
- Intra-Assay: Mean 10.3%
- Spike Recovery:
- Culture Media - 94%
- Sodium Heparin Plasma - 104%
- EDTA Plasma - 97%
- Sample Volume: 100 μL
- Components:
- Antibody
- Assay buffer
- Conjugate
- GxR IgG Microtiter plate
- Standard
- Stop solution
- pNpp Substrate
- Wash buffer concentrate
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
