Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

13-cis-Retinoic acid, MP Biomedicals™

Catalog No. ICN15651883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15651883 0.25 g
ICN15651890 500 mg
ICN15651880 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15651883 Supplier MP Biomedicals, Inc Supplier No. 0215651883
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4759-48-2
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.442
MDL Number MFCD00079542
InChI Key SHGAZHPCJJPHSC-RVDNBNLISA-N
Synonym Isotretinoin
PubChem CID 131954766
IUPAC Name (2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C

Specifications

Color Orange
Assay Percent Range 0.998
Quantity 0.25 g
Formula Weight 300.4
Physical Form Powder
Chemical Name or Material 13-cis-Retinoic acid, Cellular and Molecular Pathology
EINECSNumber 225-296-0
Recommended Storage Store at -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.