Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

13-cis-Retinoic acid, MP Biomedicals™

Catalog No. p-4405227
Click to view available options
Quantity:
0.1 g
0.25 g
500 mg

Chemical Identifiers

CAS 4759-48-2
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.442
MDL Number MFCD00079542
InChI Key SHGAZHPCJJPHSC-RVDNBNLISA-N
Synonym Isotretinoin
PubChem CID 131954766
IUPAC Name (2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C

Specifications

Color Orange
Assay Percent Range 0.998
Quantity 500 mg
Formula Weight 300.4
Physical Form Powder
Chemical Name or Material 13-cis-Retinoic acid, Cellular and Molecular Pathology
EINECSNumber 225-296-0
Recommended Storage Store at -20°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.