Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

13-cis-Retinoic Acid 97.0+%, TCI America™
SDP

Catalog No. p-7135689
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 4759-48-2
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.442
MDL Number MFCD00079542
InChI Key SHGAZHPCJJPHSC-RVDNBNLISA-N
Synonym (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, Isotretinoin
PubChem CID 131954766
IUPAC Name (2Z,4E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
SMILES CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CC(=O)O)C)C

Specifications

Melting Point 175°C
Quantity 100 mg
Formula Weight 300.44
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 13-cis-Retinoic Acid
RTECSNumber VH6440000
TSCA Yes
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.