Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter

Catalog No. 13301152
Click to view available options
Type:
Chlorine Colorimeter
Chlorine Primary Standard Kit
Chlorine Secondary Standards
Short Path Length Plastic Vial

Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.

The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.

  • Convenient scroll-driven menu system
  • Automatic switch-off
  • 16 point internal automatic data storage with date/time stamp
  • Easy to read, backlit display
  • One time zero function
  • Waterproof design
  • Angled, ergonomic design for easy bench top or hand-held use
  • Calibration to government or organizational standards (ex. NIST or ISO)
  • Calibration can be refined for very specific measurement ranges
  • Account for any interferences that are known to be present in the sample
  • Ability to export data to a PC
  • 2-year warranty

Warranty and Services

2 years

TRUSTED_SUSTAINABILITY

Specifications

Accuracy ±0.05 mg/L (0.02 to 1.00 mg/L); ±0.10 mg/L (1.01 to 2.00 mg/L); ±0.20 mg/L (2.01 to 3.00 mg/L); ±0.30 mg/L (3.01 to 4.00 mg/L); ±0.40 mg/L (4.01 to 8.00 mg/L)
Backlight Option Yes
Battery Life 17 hours operating time or 5,000 measurements in continuous mode when display backlight is off
Calibration User and factory calibration functions
Certifications/Compliance CE
Data Storage 16 with date/time stamp
Battery Type 4 x AAA
Display Backlit LCD
Dimensions (L x W x H) 155 x 75 x 35 mm
IP Rating IP67
Light Source LED
Product Line AQUAfast
Type Chlorine Colorimeter
For Use With (Application) Reagents: AC4P71, AC4P72, AQ250F, AQ250T; Vials: AC2V24, AC2V10; Standards: CLSK100, CLSK200
Includes Colorimeter with free and total chlorine powder packs (100 each), three 24 mm sample vials with lids, field case, batteries and literature
Packaging Powder packs, bulk powder dispensers
Range Low Range: 0.02 to 2.00 mg/L; High Range: 0.10 to 8.00 mg/L
Test Method Dedicated Chlorine Colorimeter
Warranty 2 Years
Wavelength 530 nm
Wavelength Accuracy ± 1 nm
Weight (Metric) 260 g (With batteries)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.