Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | 14-3-3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 2-4ug/ml, Simple Western 1:50 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227202SS
![]() |
Novus Biologicals
NBP227202SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227202
![]() |
Novus Biologicals
NBP227202 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
14-3-3 Polyclonal specifically detects 14-3-3 in Human, Mouse, Rat, Porcine, Canine, Chicken, Chinese Hamster, Feline, Opossum, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence.Specifications
14-3-3 | |
Western Blot 2-4ug/ml, Simple Western 1:50 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Pig, Canine, Chicken, Chinese Hamster, Feline, Opossum, Other, Primate | |
P27348 | |
10971 | |
A portion of amino acids 125-175 from human 14-3-3 was used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
YWHAQ | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, panda, dog, Chinese hamster, chicken, opossum, platypus, pig, frog, cat, orangutan, crab-eating macaque, and rhesus monkey, 94% in Carolina anole, cow, zebrafish, and 89% homologous in frog, and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title