Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 Antibody (SR1457), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32169450UL
Description
14-3-3 Monoclonal antibody specifically detects 14-3-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunohistochemistrySpecifications
14-3-3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry | |
SR1457 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200 | |
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
A synthesized peptide derived from human 14-3-3 (Uniprot #: P31946, P63104) | |
50 μg | |
Alzheimers Research, Cancer, Cell Biology, Cell Cycle and Replication, Growth and Development, Neuronal Cell Markers, Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
10971 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction