Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 beta Antibody (03), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306516JF549
Description
14-3-3 beta Monoclonal antibody specifically detects 14-3-3 beta in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
14-3-3 beta | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
7529 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, ELISA, Immunofluorescence | |
03 | |
Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 14-3-3 beta (NP_003395.1; Met1-Asn246). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction