Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 beta Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298453JF549
Description
14-3-3 beta Polyclonal antibody specifically detects 14-3-3 beta in Cynomolgus Monkey samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
14-3-3 beta | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha | |
Produced in rabbits immunized with purified, recombinant Cynomolgus 14-3-3 beta (Uniprot#: Q4R572-2; Met2-Asn244). | |
0.1 mL | |
Primary | |
Cynomolgus Monkey | |
Purified |
Western Blot, Immunoprecipitation | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
7529 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction