Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 beta/epsilon/zeta, Mouse anti-Human,Mouse,Rat, Alexa Fluor 750, Clone: 3C8, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197885AF750
Description
14-3-3 beta/epsilon/zeta Monoclonal specifically detects 14-3-3 beta/epsilon/zeta in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
14-3-3 beta/epsilon/zeta | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Recombinant human 14-3-3 beta. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Western Blot | |
3C8 | |
Western Blot | |
Mouse | |
Protein A purified | |
RUO | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction