Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 epsilon Antibody (CPTC-YWHAE-1), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279944V
Description
14-3-3 epsilon Monoclonal specifically detects 14-3-3 epsilon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
14-3-3 epsilon | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
YWHAE | |
Recombinant human full-length 14-3-3 epsilon protein (Uniprot: P62268) | |
0.1 mL | |
Cancer, Lipid and Metabolism | |
7531 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CPTC-YWHAE-1 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction