Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 epsilon Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297726C
Description
14-3-3 epsilon Polyclonal specifically detects 14-3-3 epsilon in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
14-3-3 epsilon | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence | |
DyLight 650 | |
14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
Produced in rabbits immunized with E. coli-derived Human 14-3-3 epsilon fragment. | |
0.1 mL | |
Cancer, Lipid and Metabolism | |
7531 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction