Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 14-3-3 epsilon Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581127
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581127 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581127 Supplier Invitrogen™ Supplier No. PA581127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human 14-3-3 epsilon/YWHAE.

14-3-3 epsilon belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the mouse ortholog. It interacts with CDC25 phosphatases, RAF1 and IRS1 proteins, suggesting its role in diverse biochemical activities related to signal transduction, such as cell division and regulation of insulin sensitivity. It has also been implicated in the pathogenesis of small cell lung cancer. Two transcript variants, one protein-coding and the other non-protein-coding, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 epsilon
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene YWHAE
Gene Accession No. P62258
Gene Alias 14-3-3 epsilon; 14-3-3 protein epsilon; 14-3-3E; AU019196; epididymis luminal protein 2; HEL2; KCIP-1; MDCR; MDS; mitochondrial import stimulation factor (MSF) L subunit; mitochondrial import stimulation factor L subunit; MSF L; protein kinase C inhibitor protein-1; tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activatioprotein epsilon polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activatioprotein, epsilon polypeptide; Ywhae
Gene Symbols YWHAE
Host Species Rabbit
Immunogen Recombinant Human 14-3-3 epsilon/YWHAE Protein (Met1-Gln255).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.