Learn More
14-3-3 epsilon Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 116482AP150UL

Description
This antibody was raised against full-length 14-3-3 epsilon.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the mouse ortholog. It interacts with CDC25 phosphatases, RAF1 and IRS1 proteins, suggesting its role in diverse biochemical activities related to signal transduction, such as cell division and regulation of insulin sensitivity. It has also been implicated in the pathogenesis of small cell lung cancer. Two transcript variants, one protein-coding and the other non-protein-coding, have been found for this gene.Specifications
14-3-3 epsilon | |
Polyclonal | |
Unconjugated | |
YWHAE | |
14 3 3 protein epsilon, 14 3 3E, 14-3-3, 14-3-3 epsilon, KCIP 1, MDCR, MDS, YWHAE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22627, 29753, 7531 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62258, P62259, P62260 | |
YWHAE | |
14-3-3 epsilon Fusion Protein Ag2247 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.