Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 eta Antibody (3G12), DyLight 550, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP192691R

Catalog No. NBP192691R


Only null left
Add to Cart

Description

Description

14-3-3 eta Monoclonal specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

14-3-3 eta
Monoclonal
DyLight 550
50 mM sodium borate with 0.05% sodium azide
YWHAH
Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917].
Primary
Human, Mouse, Rat, Bovine, Canine, Equine
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
3G12
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta
Mouse
0.1 mL
7533
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.