Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 eta Antibody (3G12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192691F
Description
14-3-3 eta Monoclonal specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.Specifications
| 14-3-3 eta | |
| Monoclonal | |
| 1mg/mL | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta | |
| Mouse | |
| 28 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Neuroscience | |
| 7533 | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| 3G12 | |
| FITC | |
| Q04917 | |
| YWHAH | |
| Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction