Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 eta Antibody (3G12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP192691MFV500
Description
14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, KnockDownSpecifications
14-3-3 eta | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Canine | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, KnockDown | |
3G12 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Knockout Validated | |
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta | |
Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917]. | |
0.1 mL | |
Apoptosis, Cancer, Neuroscience | |
7533 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction