Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 eta Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299642AF700
Description
14-3-3 eta Polyclonal specifically detects 14-3-3 eta in Human samples. It is validated for Western Blot, ELISA.Specifications
14-3-3 eta | |
Polyclonal | |
Western Blot, ELISA | |
14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta | |
Produced in rabbits immunized with purified, recombinant 14-3-3 eta (Q04917; Gly2-Asn246). | |
0.1 mL | |
Apoptosis, Cancer, Neuroscience | |
7533 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction