Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 eta Monoclonal Antibody (3G12)
14-3-3 eta Monoclonal Antibody (3G12)
Supplier: Thermo Scientific MA524792
Description
14-3-3 eta Monoclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
14-3-3 eta | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 5mM sodium azide | |
P68509, P68510, P68511, Q04917 | |
YWHAH | |
Full length recombinant human 14-3-3 eta. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Canine, Bovine, Horse | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
3G12 | |
Unconjugated | |
YWHAH | |
14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta-subtype; 14-3-3e; 1433F; fb93e02; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; wu:fb51h04; wu:fb93e02; YWHA1; YWHA114-3-3 eta; ywhag; YWHAH; zgc:63883; zgc:63883 protein | |
Mouse | |
Affinity chromatography | |
RUO | |
100062878, 100687866, 22629, 25576, 282126, 7533 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction