Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 14-3-3 eta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118366
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118366 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5118366 Supplier Invitrogen™ Supplier No. PA5118366
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

14-3-3 eta Polyclonal Antibody for Western Blot, IP

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and bovine orthologs. This gene contains a 7 bp repeat sequence in its 5' UTR, and changes in the number of this repeat has been associated with early-onset schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 eta
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene YWHAH
Gene Accession No. Q04917
Gene Alias 14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta-subtype; 14-3-3e; 1433F; fb93e02; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; wu:fb51h04; wu:fb93e02; YWHA1; YWHA114-3-3 eta; ywhag; YWHAH; zgc:63883; zgc:63883 protein
Gene Symbols YWHAH
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the center region of the Human 14-3-3 eta/YWHAH.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7533
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.