Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 gamma Antibody (13C9), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32624650UL
Description
14-3-3 gamma Monoclonal antibody specifically detects 14-3-3 gamma in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
14-3-3 gamma | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
13C9 | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
A synthesized peptide derived from Human 14-3-3 gamma [UniProt P61981] | |
50 μL | |
Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
7532 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction