Learn More
14-3-3 GAMMA Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 123811AP150UL

Description
This antibody specifically recognizes gamma isoform of 14-3-3.
14-3-3 gamma encodes a product that belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 100% identical to the rat ortholog. It is induced by growth factors in human vascular smooth muscle cells, and is also highly expressed in skeletal and heart muscles, suggesting an important role for this protein in muscle tissue. It has been shown to interact with RAF1 and protein kinase C, proteins involved in various signal transduction pathways.Specifications
14-3-3 GAMMA | |
Polyclonal | |
Unconjugated | |
YWHAG | |
14 3 3 protein gamma, 14 3 3GAMMA, 14-3-3, 14-3-3 GAMMA, KCIP 1, YWHAG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22628, 489818, 56010, 7532 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.42 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61981, P61982, P61983 | |
Ywhag | |
14-3-3 GAMMA-Specific Fusion Protein Ag3047 | |
150 μL | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.