Learn More
14-3-3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E10A3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | 14-3-3 |
---|---|
Clone | 2E10A3 |
Concentration | 0.71 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6500
![]() ![]() |
Proteintech Group Inc
660611IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
66061-1-Ig recognizes 14-3-3 tau (YWHAQ), eta (YWHAH), and zeta (YWHAZ), but not epsilon (YWHAE).
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5′ UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.Specifications
14-3-3 | |
0.71 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
14 3 3, 14 3 3 protein T cell, 14 3 3 protein tau, 14 3 3 protein theta, 14-3-3 tau, HS1, Protein HS1, YWHAQ | |
Ywhaq | |
IgG1 | |
Protein G | |
Antibody |
2E10A3 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P27348, P68254, P68255 | |
100048964, 10971, 22630, 25577 | |
14-3-3 Fusion Protein Ag6055 | |
Primary | |
-20°C | |
YWHAQ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.