Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Pan Monoclonal Antibody (CG15), Invitrogen™

Catalog No. PIMA512242
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512242 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512242 Supplier Thermo Scientific Supplier No. MA512242

Mouse Monoclonal Antibody

MA5-12242 targets 14.3.3 pan in IP, ICC/IF, IHC (P) and WB applications and shows reactivity with Human and Mouse samples. The MA5-12242 immunogen is 14-3-3 gamma recombinant human protein.

At least seven isoforms comprise the highly conserved 14-3-3 family of homo- and heterodimeric proteins that are abundantly expressed in all eukaryotic cells. Although more than seven isoforms of 14-3-3 have been described, some redundancies have appeared upon sequencing. The 14-3-3s are thought to be key regulators of signal transduction events mediated through their binding to serine-phosphorylated proteins. By interacting with Cdc25C, 14-3-3 regulates entry into the cell cycle and, through interaction with Bad, prevents apoptosis. Other proteins that have been shown to bind to 14-3-3s include members of the protein kinase C family, Cbl, IRS-1, polyoma middle-T antigen, nitrate reductase, S-raf and the IGF-1 receptor. Detection of 14-3-3 proteins in cerebrospinal fluid has been shown to be quite useful in the differential diagnosis of Creutzfeldt-Jakob disease and other prion diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 Pan
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone CG15
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene YWHAG
Gene Accession No. O70456, P27348, P31946, P31947, P61981, P61982, P62258, P62259, P63101, P63104, P68254, Q9CQV8
Gene Alias 1110013I11Rik; 1300003C17Rik; 14-3-3; 14-3-3 alpha; 14-3-3 beta; 14-3-3 delta; 14-3-3 epsilon; 14-3-3 gamma; 14-3-3 protein beta; 14-3-3 protein beta/alpha; 14-3-3 protein beta/alpha, N-terminally processed; 14-3-3 protein beta/alpha-A; 14-3-3 protein beta/alpha-B; 14-3-3 protein beta-subtype; 14-3-3 protein epsilon; 14-3-3 protein gamma; 14-3-3 protein gamma subtype; 14-3-3 protein gamma, N-terminally processed; 14-3-3 protein gamma-A; 14-3-3 protein gamma-B; 14-3-3 protein gamma-subtype; 14-3-3 protein sigma; 14-3-3 protein tau; 14-3-3 protein T-cell; 14-3-3 protein theta; 14-3-3 protein zeta; 14-3-3 protein zeta/delta; 14-3-3 protein/cytosolic phospholipase A2; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3 tau; 14-3-3 theta; 14-3-3 zeta; 14-3-3beta; 14-3-3d; 14-3-3E; 14-3-3G; 14-3-3gamma; 14-3-3-like protein; 14-3-3t; 14-3-3z; 14-3-3zeta; 14-3-3-zeta; 1C5; 2700028P07Rik; 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; 3-monooxgenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; 3-monooxygenase/tryptophan 5 monooxygenase activation protein beta polypeptide; 3-monooxygenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide; AA409740; AI596267; AL022924; AU019196; AU020854; AU021156; brain protein 14-3-3, beta isoform; D7Bwg1348e; epididymis luminal protein 2; epididymis luminal protein 4; epididymis secretory protein Li 1; epididymis secretory protein Li 3; epididymis secretory protein Li 93; epithelial cell marker protein 1; Er; factor activating exoenzyme S; FAS; GW128; HEL2; HEL4; HEL-S-1; HEL-S-3; HEL-S-93; HME1; HS1; KCIP-1; MDCR; MDS; mitochondrial import stimulation factor (MSF) L subunit; mitochondrial import stimulation factor L subunit; Mitochondrial import stimulation factor S1 subunit; Mkrn3; Mme1; MSF L; Msfs1; phospholipase A2; PPP1R170; prepronerve growth factor RNH-1; Protein 1054; Protein HS1; Protein kinase C inhibitor protein 1; protein kinase C inhibitor protein-1; protein phosphatase 1, regulatory subunit 170; protein tau; protein, theta; R74690; SEZ-2; SFN; Stratifin; tryosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein gamma; tryosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein gamma polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta polypeptide; tryosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3/tryptophan 5 -monooxygenase activation protein beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, beta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, gamma polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, theta polypeptide; tyrosine 3/tryptophan 5 -monooxygenase activation protein, zeta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5 monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, beta polypeptide; tyrosine 3-monooxgenase/tryptophan 5-monooxgenase activation protein, gamma polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta L homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta S homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma L homeolog; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein gamma S homeolog
Gene Symbols Sfn, YWHAB, YWHAE, YWHAG, YWHAQ, YWHAZ
Host Species Mouse
Immunogen 14-3-3 gamma recombinant human protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10971, 22627, 22628, 22630, 22631, 2810, 54401, 55948, 7529, 7531, 7532, 7534
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.