Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 14-3-3, Rabbit, Unlabeled, Polyclonal,

Catalog No. AB9748I Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB9748I 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9748I Supplier MilliporeSigma™ Supplier No. AB9748I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

14-3-3 protein beta/alpha (KCIP-1, Protein 1054, Protein kinase C inhibitor protein 1), epsilon (14-3-3E), eta (Protein AS1), gamma (KCIP-1, Protein kinase C inhibitor protein 1), theta (14-3-3 protein T-cell, 14-3-3 protein tau, Protein HS1), zeta/delta (KCIP-1, Protein kinase C inhibitor protein 1) are encoded by the YWHAB, YWHAE, YWHAH, YWHAG, YWHAQ, YWHAZ gene (Gene ID 7529, 7531, 7533, 7532, 10971, 7534, respectively) in human. Mammals express seven distinct 14-3-3 isoforms (gamma, epsilon, beta, zeta, sigma, theta, tau) that form multiple homo- and hetero- dimmers, thus rendering 14-3-3 as a key multifunctional regulatory molecule. 14-3-3 isomers have been implicated in several neurological disorders: for example isoforms gamma, epsilon and beta are expressed in the cerebrospinal fluid of Creutzfeldt-Jakob patients, but not patients suffering from other neurological conditions such as Alzheimer's. The 14-3-3 pan antibody is one of a panel of antibodies that were generated against 14-3-3 isoforms and have demonstrated their potential for use as an ELISA-based diagnostic tool. These antibodies will allow the research to investigate the role of this protein in key cellular processes and neurological disorders such as Creutzfeldt-Jakob, Alzheimer's and Parkinson's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene YWHAB, YWHAE, YWHAG, YWHAZ, YWHA1, YWHAH, YWHAQ
Gene Accession No. P31946;P63104;P62258;Q04917;P61981;P27348
Host Species Rabbit
Immunogen A mixture of KLH-conjugated linear peptides corresponding the the N-terminal end sequences of human 14-3-3 alpha/beta (YWHAB), gamma (YWHAG), zeta/delta (YWHAZ), epsilon (YWHAE), eta (YWHAH or YWHA1), and theta (YWHAQ).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_003395;NP_003397;NP_006752;NP_003396;NP_036611;NP_006817
Test Specificity This antibody targets the N-terminal end sequences of 14-3-3 alpha/beta (YWHAB), gamma (YWHAG), zeta/delta (YWHAZ), epsilon (YWHAE), eta (YWHAH or YWHA1), and theta (YWHAQ).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.