Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 14-3-3 sigma Monoclonal Antibody (02)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA535997
Description
14-3-3 sigma Monoclonal Antibody for Flow
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
Specifications
14-3-3 sigma | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
P31947 | |
SFN | |
Recombinant Human 14-3-3 sigma/Stratifin/YWHAS Protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
02 | |
Unconjugated | |
SFN | |
14-3-3 protein sigma; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3sigma; epithelial cell marker protein 1; Er; HGNC:10773; HME1; Mkrn3; Mme1; SFN; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; Ywhas | |
Mouse | |
Protein A | |
RUO | |
2810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction