Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 14-3-3 sigma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587755
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587755 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587755 Supplier Invitrogen™ Supplier No. PA587755
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MERASLIQKA KLAEQAERYE DMAAFMKGAV EKGEELSCEE RNLLSVAYKN VVGGQRAAWR VLSSIEQKSN EEGSEEKGPE VREYREKVET ELQGVCDTVL Positive Samples: HT-29, MCF7; Cellular Location: Cytoplasm, Nucleus.

This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 sigma
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SFN
Gene Accession No. O70456, P31947
Gene Alias 14-3-3 protein sigma; 14-3-3 sigma; 14-3-3 sigma protein; 14-3-3sigma; epithelial cell marker protein 1; Er; HGNC:10773; HME1; Mkrn3; Mme1; SFN; Stratifin; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, sigma polypeptide; Ywhas
Gene Symbols SFN
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human 14-3-3 sigma (NP_0061331).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2810, 313017, 55948
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.