Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

14-3-3 Sigma Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501571937
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1937 100 μL
50-157-1938 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1937 Supplier Bethyl Laboratories, Inc Supplier No. IHC00346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Crab-eating macaque and Wh

14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to <FONT face=Symbol>g</FONT>-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5′ CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.

Specifications

Antigen 14-3-3 Sigma
Applications Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene SFN
Gene Accession No. P31947
Gene Alias 14-3-3 protein sigma, 14-3-3 sigma, epithelial cell marker protein 1
Gene Symbols SFN
Host Species Rabbit
Immunogen Between 198 and 248
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2810
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.