Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 sigma/Stratifin Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299631APC
Description
14-3-3 sigma/Stratifin Polyclonal antibody specifically detects 14-3-3 sigma/Stratifin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
14-3-3 sigma/Stratifin | |
Polyclonal | |
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS | |
Produced in rabbits immunized with purified, recombinant Human 14-3-3 sigma/Stratifin (Accession#: NP_006133.1; Met1-Ser248). | |
0.1 mL | |
Cell Cycle and Replication, Phospho Specific | |
2810 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction