Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 tau/theta Antibody (1A1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP121301PE
Description
14-3-3 tau/theta Monoclonal specifically detects 14-3-3 tau/theta in Human samples. It is validated for Flow Cytometry.Specifications
14-3-3 tau/theta | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
YWHAQ | |
Recombinant human 14-3-3 tau/theta (1-245aa) purified from E. coli (NP_006817). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry | |
1A1 | |
Flow Cytometry | |
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide | |
Mouse | |
Protein A or G purified | |
RUO | |
10971 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction