Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 14-3-3 theta Recombinant Rabbit Monoclonal Antibody (SD084-02)
GREENER_CHOICE

Catalog No. PIMA532417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532417 Supplier Invitrogen™ Supplier No. MA532417
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5' UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 14-3-3 theta
Applications Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone SD084-02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene YWHAQ
Gene Accession No. P27348, P68254, P68255
Gene Alias 14-3-3; 14-3-3 protein tau; 14-3-3 protein T-cell; 14-3-3 protein theta; 14-3-3 tau; 14-3-3 theta; 14-3-3t; 1C5; 2700028P07Rik; AA409740; AU021156; HS1; OTTHUMP00000165887; Protein HS1; protein tau; protein, theta; R74690; tyrosine 3/tryptophan 5 -monooxygenase activation protein, theta polypeptide; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide; Unknown (protein for MGC:127486); YWHAQ
Gene Symbols YWHAQ
Host Species Rabbit
Immunogen Synthetic peptide within Human 14-3-3 Theta aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10971, 22630, 25577
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.