Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
14-3-3 zeta/beta Rabbit anti-Human, Mouse, Rat, Clone: 3J3J9, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31676920UL
This item is not returnable.
View return policy
Description
14-3-3 zeta/beta Monoclonal antibody specifically detects 14-3-3 zeta/beta in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| 14-3-3 zeta/beta | |
| Monoclonal | |
| Unconjugated | |
| KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, zeta polypeptide | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human 14-3-3 zeta/beta (P31946). | |
| 20 μg | |
| Cell Cycle and Replication | |
| 7534 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| 3J3J9 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction