Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-Crown-5, 98%

Catalog No. AAA1226514
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 33100-27-5 | C10H20O5 | 220.265 g/mol

15-Crown-5 is used as an efficient phase transfer catalyst and as a complexing agent. It is used to isolate oxonium ion (H7O3)+ salts especially from a solution of tetrchloroauric acid. It catalyzes the O-alkylation of the sodium salts of carboxylic acids in the penicillin and cephalosporin series, thereby facilitating esterification reaction without usage of acid. It is also used to facilitate the Williamson synthesis of ethers with hindered alcohols and sodium hydride. It is used with lithium aluminum hydride, for performing reduction reactions in hydrocarbon solvents. Further, it is involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
15-Crown-5 is used as an efficient phase transfer catalyst and as a complexing agent. It is used to isolate oxonium ion (H7O3)+ salts especially from a solution of tetrchloroauric acid. It catalyzes the O-alkylation of the sodium salts of carboxylic acids in the penicillin and cephalosporin series, thereby facilitating esterification reaction without usage of acid. It is also used to facilitate the Williamson synthesis of ethers with hindered alcohols and sodium hydride. It is used with lithium aluminum hydride, for performing reduction reactions in hydrocarbon solvents. Further, it is involved in the Horner-Wadsworth-Emmons reaction to prepare stilbenes from aldehydes.

Solubility
Miscible with organic solvents.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33100-27-5
Molecular Formula C10H20O5
Molecular Weight (g/mol) 220.265
MDL Number MFCD00005110
InChI Key VFTFKUDGYRBSAL-UHFFFAOYSA-N
Synonym 15-crown-5, 15-crown-5 ether, 15-crown 5-ether, 1,4,10,13-pentaoxacyclopentadecane, ccris 3586, 15-crown 5, crown ether 15-crown-5, 5-19-12-00252 beilstein handbook reference, ksc917c1b
PubChem CID 36336
ChEBI CHEBI:32401
IUPAC Name 1,4,7,10,13-pentaoxacyclopentadecane
SMILES C1COCCOCCOCCOCCO1

Specifications

Density 1.11
Boiling Point 150°C to 155°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.465
Quantity 25 g
Beilstein 1618144
Sensitivity Hygroscopic
Solubility Information Miscible with organic solvents.
Formula Weight 220.27
Percent Purity 98%
Chemical Name or Material 15-Crown-5
Show More Show Less
Hazard Category H302+H332-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-379-6
RTECSNumber SB0200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.