Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ 15-deoxy-Delta 12, 14-PGJ2 ELISA Kit (Colorimetric)
SDP

Catalog No. NB2612855KT Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
5 Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB2612855KT 5 Kit
NBP261285 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB2612855KT Supplier Novus Biologicals™ Supplier No. NBP2612855KIT
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

The Non-species specific 15-deoxy-Delta 12, 14-PGJ2 ELISA Kit (Colorimetric) measures non-species specific 15-deoxy-Delta 12, 14-PGJ2 in plasma, serum, and urine from any species.

Specifications

For Use With (Application) ELISA
Quantity 5 Kit
Storage Requirements Store at 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.