Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-Lipoxygenase 1 Antibody (OTI3G8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270169H
Description
15-Lipoxygenase 1 Monoclonal specifically detects 15-Lipoxygenase 1 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
15-Lipoxygenase 1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ALOX15 | |
Full length recombinant protein of human 15-Lipoxygenase 1 (NP_001131) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat, Canine, Primate, Monkey | |
IgG2b |
Western Blot, Immunohistochemistry | |
OTI3G8 | |
Western Blot, Immunohistochemistry | |
15-lipooxygenase-1, 15-LOX-1, 15LOX-1, arachidonate 15-lipoxygenase, Arachidonate omega-6 lipoxygenase, EC 1.13.11, EC 1.13.11.33, LOG15,15-LOX | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
246.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction