Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
15-Lipoxygenase 2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300321MFV500
Description
15-Lipoxygenase 2 Polyclonal antibody specifically detects 15-Lipoxygenase 2 in Human samples. It is validated for ELISASpecifications
15-Lipoxygenase 2 | |
Polyclonal | |
ELISA | |
15-lipoxygenase 2, 15-LOX-B, arachidonate 15-lipoxygenase 2, arachidonate 15-lipoxygenase B, Arachidonate 15-lipoxygenase type II, arachidonate 15-lipoxygenase, second type, arachidonate 15-lipoxygenase, type B, arachidonate omega(6) lipoxygenase, EC 1.13.11, EC 1.13.11.33,15-LOX-215S-lipoxygenase | |
Produced in rabbits immunized with purified, recombinant Human 15-Lipoxygenase 2 (Accession#: AAH35217.1; Met1-Ile676). | |
0.1 mL | |
Lipid and Metabolism | |
247 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction