Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15 Lipoxygenase 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen 15 Lipoxygenase 2
Concentration 0.13 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-8467
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
130731AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

13073-1-AP can also recognize the mouse Alox8 protein.

ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.
Specifications

Specifications

15 Lipoxygenase 2
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
O15296, O35936, Q8K4F2
11688, 247, 266604
15 Lipoxygenase 2 Fusion Protein Ag3741
Primary
-20°C
ALOX15B
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
15 Lipoxygenase 2, 15 LOX 2, 15 LOX B, ALOX15B, ALOX8, Arachidonate 15 lipoxygenase B
Alox15b, Alox8
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.