Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

15-Lox-2 Polyclonal Antibody, Abbomax

Catalog No. 502203910
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3910 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3910 Supplier Abbomax Inc Supplier No. 600590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HT-29 cells Cellular location: Cytoplasmic.

15-Lox-2 Polyclonal antibody specifically detects 15-Lox-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen 15-Lox-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene ALOX15B
Gene Accession No. O15296, O35936, Q8K4F2
Gene Alias 15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; 8S-LOX; ALOX15B; Alox8; arachidonate 15-lipoxygenase 2; arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate 15-lipoxygenase, type B; arachidonate 8-lipoxygenase; arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; linoleate 13-lipoxygenase 15-LOb
Host Species Rabbit
Immunogen A synthetic peptide LP YTY LDP PLI ENS VSI corresponding to the internal portion of human LOX2 protein sequence
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11688, 247, 266604
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.